Assay Details
Target Gene Details
Entrez Gene ID: | 57150 |
Gene Name: | small integral membrane protein 8 |
Gene Aliases: |
C6orf162, dJ102H19.2 |
Location: |
Chr.6:87322588-87342328 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMIM8 | NM_001042493.2 | 4 | 2139 | NP_001035958.1 |
| NM_001287445.1 | 3 | 1008 | NP_001274374.1 | |
| NM_020425.5 | 3 | 2118 | NP_065158.3 | |
| AI239454.1 | 1 | 299 | ||
| AI681117.1 | 1 | 303 | ||
| AJ420558.1 | 1 | 1314 |

Genomic Map