Assay Details
Target Gene Details
Entrez Gene ID: | 55147 |
Gene Name: | RNA binding motif protein 23 |
Gene Aliases: |
CAPERbeta, PP239, RNPC4 |
Location: |
Chr.14:22900645-22919187 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
More Information
Set Membership: |
|

Genomic Map