Assay Details
Target Gene Details
Entrez Gene ID: | 54476 |
Gene Name: | ring finger protein 216 |
Gene Aliases: |
CAHH, TRIAD3, U7I1, UBCE7IP1, ZIN |
Location: |
Chr.7:5620041-5781730 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF216 | NM_207111.3 | 17 | 4059 | NP_996994.1 |
| NM_207116.2 | 17 | 3888 | NP_996999.1 | |
| XM_005249785.2 | 19 | 4379 | XP_005249842.1 | |
| XM_011515436.1 | 15 | 3798 | XP_011513738.1 | |
| XM_017012363.1 | 17 | 3743 | XP_016867852.1 | |
| XM_017012365.1 | 16 | 3701 | XP_016867854.1 | |
| AK024787.1 | 3 | 1631 | ||
| BC000787.2 | 11 | 2473 | AAH00787.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830890 | Chr.7:5425302 - 5665527 on Build GRCh38 | Loss |
|
| nsv436904 | Chr.7:5035574 - 6713282 on Build GRCh38 | Insertion |
|
| nsv1034883 | Chr.7:5612049 - 5656516 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map