Assay Details
Target Gene Details
Entrez Gene ID: | 64921 |
Gene Name: | CAS1 domain containing 1 |
Gene Aliases: |
C7orf12, NBLA04196 |
Location: |
Chr.7:94509858-94633465 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASD1 | NM_022900.4 | 7 | 799 | NP_075051.4 |
| XM_006716092.3 | 5 | 294 | XP_006716155.1 | |
| XM_006716093.3 | 7 | 637 | XP_006716156.2 | |
| XM_011516495.2 | 7 | 637 | XP_011514797.1 | |
| XM_017012546.1 | 7 | 637 | XP_016868035.1 | |
| XM_017012547.1 | 8 | 795 | XP_016868036.1 | |
| AF355594.1 | 7 | 592 | AAL33538.1 | |
| AF397424.2 | 7 | 719 | AAK97479.2 | |
| AK123866.1 | 7 | 637 | ||
| AK314329.1 | 7 | 597 | ||
| BC063284.1 | 7 | 784 | AAH63284.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1023193 | Chr.7:93602376 - 95828660 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map