Assay Details
Target Gene Details
Entrez Gene ID: | 10105 |
Gene Name: | peptidylprolyl isomerase F |
Gene Aliases: |
CYP3, CyP-M, Cyp-D, CypD |
Location: |
Chr.10:79347464-79355334 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPIF | NM_005729.3 | 6 | 869 | NP_005720.1 |
| XM_005269379.2 | 6 | 929 | XP_005269436.1 | |
| AK296669.1 | 5 | 786 | ||
| AL832720.1 | 1 | 3867 | ||
| AY358854.1 | 1 | 1004 | AAQ89213.1 | |
| BC005020.1 | 6 | 855 | AAH05020.1 | |
| BC110299.1 | 5 | 770 | AAI10300.1 | |
| M80254.1 | AAA58434.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161981 | Chr.10:79309656 - 79385886 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map