Assay Details
Target Gene Details
Entrez Gene ID: | 146853 |
Gene Name: | chromosome 17 open reading frame 50 |
Gene Aliases: |
- |
Location: |
Chr.17:35760897-35765079 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C17orf50 | NM_145272.3 | 3 | 497 | NP_660315.2 |
| AY557347.1 | 2 | 452 | AAS67921.1 | |
| DB452451.1 | 3 | 408 |
Target Gene Details
Entrez Gene ID: | 79148 |
Gene Name: | matrix metallopeptidase 28 |
Gene Aliases: |
EPILYSIN, MM28, MMP-25, MMP-28, MMP25 |
Location: |
Chr.17:35756249-35795707 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP28 | NR_111988.1 | |||
| XM_011525231.1 | XP_011523533.1 | |||
| XM_017025064.1 | XP_016880553.1 | |||
| AK122604.1 | ||||
| BM702847.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map