Assay Details
Target Gene Details
Entrez Gene ID: | 23078 |
Gene Name: | von Willebrand factor A domain containing 8 |
Gene Aliases: |
KIAA0564 |
Location: |
Chr.13:41566825-41961111 on Build GRCh38 |
Assay Gene Location: | Within Exon 52 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWA8 | NM_015058.1 | 45 | 5778 | NP_055873.1 |
| XM_006719791.3 | 37 | 4734 | XP_006719854.1 | |
| XM_017020469.1 | 47 | 5837 | XP_016875958.1 | |
| XM_017020472.1 | 29 | 3766 | XP_016875961.1 | |
| XM_017020473.1 | 28 | 3659 | XP_016875962.1 | |
| AB011136.1 | 34 | 4318 | BAA25490.1 | |
| AK025432.1 | 5 | 720 | ||
| AK308568.1 | 4 | 503 | ||
| BC131802.1 | 46 | 5611 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1634n100 | Chr.13:41408612 - 41936985 on Build GRCh38 | Gain |
|
| nsv455865 | Chr.13:41522908 - 41951740 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map