Assay Details
Target Gene Details
Entrez Gene ID: | 79676 |
Gene Name: | 2-oxoglutarate and iron dependent oxygenase domain containing 2 |
Gene Aliases: |
- |
Location: |
Chr.12:122974703-122980041 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGFOD2 | NM_001304833.1 | 5 | 492 | NP_001291762.1 |
| NM_001304834.1 | 6 | 923 | NP_001291763.1 | |
| NM_001304835.1 | 5 | 1043 | NP_001291764.1 | |
| NM_001304836.1 | 5 | 812 | NP_001291765.1 | |
| NM_001304837.1 | 4 | 673 | NP_001291766.1 | |
| NM_001304838.1 | 5 | 730 | NP_001291767.1 | |
| NM_024623.2 | 6 | 879 | NP_078899.1 | |
| AK023553.1 | 6 | 775 | BAB14610.1 | |
| AK094820.1 | 5 | 475 | ||
| AK095481.1 | 4 | 581 | ||
| BC098119.1 | 6 | 655 | AAH98119.1 | |
| BC098293.1 | 6 | 655 | AAH98293.1 | |
| BC101940.1 | 5 | 462 | AAI01941.1 | |
| BC105637.1 | 5 | 462 | AAI05638.1 | |
| BX640675.1 | 5 | 464 | CAE45807.1 | |
| BX640734.1 | 6 | 523 | CAE45849.1 | |
| CR457316.1 | 3 | 263 | CAG33597.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv914 | Chr.12:122977362 - 123008698 on Build GRCh38 | Insertion |
|
| dgv22n68 | Chr.12:122911380 - 123104743 on Build GRCh38 | Loss |
|
| nsv455732 | Chr.12:122943026 - 123090291 on Build GRCh38 | Loss |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map