Assay Details
Target Gene Details
Entrez Gene ID: | 10899 |
Gene Name: | jumping translocation breakpoint |
Gene Aliases: |
HJTB, HSPC222, PAR, hJT |
Location: |
Chr.1:153974269-153977975 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JTB | NM_006694.3 | 1 | 82 | NP_006685.1 |
| BU527541.1 | 1 | 97 |
More Information
Set Membership: |
|

Genomic Map