Assay Details
Target Gene Details
Entrez Gene ID: | 23647 |
Gene Name: | ADP ribosylation factor interacting protein 2 |
Gene Aliases: |
POR1 |
Location: |
Chr.11:6474683-6481479 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARFIP2 | NM_001242854.1 | NP_001229783.1 | ||
| NM_001242855.1 | NP_001229784.1 | |||
| NM_001242856.1 | NP_001229785.1 | |||
| NM_012402.3 | NP_036534.1 | |||
| XM_005252840.4 | XP_005252897.1 | |||
| AK026390.1 | ||||
| AK300855.1 | ||||
| AK301856.1 | ||||
| AK301972.1 | ||||
| AK304512.1 | ||||
| AK309198.1 | 2 | 246 | ||
| AK310833.1 | ||||
| AK311086.1 | ||||
| AK311655.1 | 3 | 219 | ||
| BC000392.2 | AAH00392.1 | |||
| DB099222.1 | ||||
| U52522.1 | AAA97924.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv972944 | Chr.11:6478192 - 6497815 on Build GRCh38 | Duplication |
|
| esv3625266 | Chr.11:6460034 - 6490367 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76285816] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map