Assay Details
Target Gene Details
Entrez Gene ID: | 9187 |
Gene Name: | solute carrier family 24 member 1 |
Gene Aliases: |
CSNB1D, HsT17412, NCKX, NCKX1, RODX |
Location: |
Chr.15:65611405-65660995 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC24A1 | NM_001301031.1 | 1 | 926 | NP_001287960.1 |
| NM_001301032.1 | 1 | 926 | NP_001287961.1 | |
| NM_001301033.1 | 4 | 1424 | NP_001287962.1 | |
| NM_004727.2 | 2 | 1087 | NP_004718.1 | |
| XM_005254778.3 | 3 | 1061 | XP_005254835.1 | |
| XM_005254781.3 | 3 | 1061 | XP_005254838.1 | |
| XM_006720768.3 | 3 | 1061 | XP_006720831.1 | |
| XM_011522219.1 | 3 | 1155 | XP_011520521.1 | |
| XM_011522220.2 | 3 | 1061 | XP_011520522.1 | |
| XM_011522221.2 | 3 | 1061 | XP_011520523.1 | |
| XM_011522222.2 | 3 | 1061 | XP_011520524.1 | |
| XM_011522226.2 | 3 | 1061 | XP_011520528.1 | |
| XM_017022724.1 | 3 | 1061 | XP_016878213.1 | |
| XM_017022725.1 | 3 | 1061 | XP_016878214.1 | |
| AB014602.1 | 3 | 2275 | BAA31677.2 | |
| AF026132.1 | 1 | 800 | AAB97832.1 | |
| AF062921.1 | 1 | 800 | AAC16732.1 | |
| AF062922.1 | 2 | 1087 | AAC77912.1 | |
| AK304006.1 | 4 | 1424 | ||
| BC117263.1 | 1 | 882 | ||
| BC143375.1 | 1 | 882 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833042 | Chr.15:65446079 - 65656395 on Build GRCh38 | Loss |
|
| nsv820238 | Chr.15:65624239 - 65624886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map