Assay Details
Target Gene Details
Entrez Gene ID: | 11240 |
Gene Name: | peptidyl arginine deiminase 2 |
Gene Aliases: |
PAD-H19, PAD2, PDI2 |
Location: |
Chr.1:17066761-17119453 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PADI2 | NM_007365.2 | NP_031391.2 | ||
| XM_017000148.1 | XP_016855637.1 | |||
| AB023211.1 | ||||
| AB030176.1 | BAA82557.1 | |||
| AK127006.1 | ||||
| AK295783.1 | ||||
| BC009701.1 | AAH09701.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758925 | Chr.1:16437838 - 17157486 on Build GRCh38 | Gain+Loss |
|
| nsv545690 | Chr.1:17040376 - 17159158 on Build GRCh38 | Gain |
|
| nsv834280 | Chr.1:16888218 - 17123006 on Build GRCh38 | Loss |
|
| nsv1145323 | Chr.1:17076286 - 17100733 on Build GRCh38 | Deletion |
|
| esv3585359 | Chr.1:17077448 - 17111045 on Build GRCh38 | Gain |
|
| esv2422314 | Chr.1:17080898 - 17216627 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map