Assay Details
Target Gene Details
Entrez Gene ID: | 10488 |
Gene Name: | cAMP responsive element binding protein 3 |
Gene Aliases: |
LUMAN, LZIP, sLZIP |
Location: |
Chr.9:35732320-35737008 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CREB3 | NM_006368.4 | NP_006359.3 | ||
| AF009368.1 | AAB69652.1 | |||
| AF029674.1 | AAB84166.1 | |||
| AF211848.1 | AAG43528.1 | |||
| AK299781.1 | ||||
| AK313635.1 | ||||
| BC009402.2 | AAH09402.1 | |||
| BC010158.2 | AAH10158.1 | |||
| BG701230.1 | ||||
| FJ263669.1 | ||||
| U88528.1 | 2 | 435 | AAC04325.1 |
More Information
Set Membership: |
|

Genomic Map