Assay Details
Target Gene Details
Entrez Gene ID: | 26280 |
Gene Name: | interleukin 1 receptor accessory protein like 2 |
Gene Aliases: |
IL-1R9, IL1R9, IL1RAPL-2, TIGIRR-1 |
Location: |
Chr.X:104566315-105769729 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL1RAPL2 | NM_017416.1 | NP_059112.1 | ||
| XM_017029403.1 | XP_016884892.1 | |||
| AF181285.1 | AAF59412.1 | |||
| AF212016.1 | AAF61307.1 | |||
| AF284436.1 | AAG21370.1 | |||
| AJ272208.1 | CAB86868.1 | |||
| AJ290436.1 | CAB89867.1 | |||
| AK290196.1 | ||||
| BC104784.1 | AAI04785.1 | |||
| BC104786.1 | AAI04787.1 |
Target Gene Details
Entrez Gene ID: | 56157 |
Gene Name: | testis expressed 13A |
Gene Aliases: |
- |
Location: |
Chr.X:105218929-105220693 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEX13A | NM_001291277.1 | 3 | 1290 | NP_001278206.1 |
| NM_031274.4 | 3 | 1316 | NP_112564.1 | |
| AF285597.1 | 3 | 1297 | AAK31976.1 | |
| BC042547.1 | 3 | 1271 | AAH42547.1 | |
| BC108734.1 | 3 | 1289 | AAI08735.1 |
More Information
Set Membership: |
|

Genomic Map