Assay Details
Target Gene Details
Entrez Gene ID: | 54862 |
Gene Name: | coiled-coil and C2 domain containing 1A |
Gene Aliases: |
FREUD-1, Freud-1/Aki1, MRT3 |
Location: |
Chr.19:13906143-13930880 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Intron 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CC2D1A | NM_017721.4 | NP_060191.3 | ||
| XM_005259972.2 | XP_005260029.1 | |||
| XM_005259973.2 | XP_005260030.1 | |||
| XM_005259974.2 | XP_005260031.1 | |||
| XM_005259975.2 | XP_005260032.1 | |||
| AB097002.1 | BAC77355.1 | |||
| AF536205.1 | AAN04488.1 | |||
| AK026371.1 | BAB15464.1 | |||
| AK315068.1 | ||||
| BC048345.1 | AAH48345.1 | |||
| BC064981.1 | AAH64981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953979 | Chr.19:13795787 - 14006888 on Build GRCh38 | Deletion |
|
| nsv833757 | Chr.19:13725904 - 13951988 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map