Assay Details
Target Gene Details
Entrez Gene ID: | 9368 |
Gene Name: | SLC9A3 regulator 1 |
Gene Aliases: |
EBP50, NHERF, NHERF-1, NHERF1, NPHLOP2 |
Location: |
Chr.17:74748612-74769360 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC9A3R1 | NM_004252.4 | 1 | 427 | NP_004243.1 |
| AF015926.1 | 1 | 404 | AAC52084.1 | |
| AF036241.1 | 1 | 362 | AAC04572.1 | |
| AK026581.1 | 1 | 410 | ||
| AK094467.1 | ||||
| AK128474.1 | 2 | 376 | ||
| AK312498.1 | 1 | 443 | ||
| BC001443.2 | 1 | 380 | AAH01443.1 | |
| BC003361.1 | 1 | 397 | AAH03361.1 | |
| BC011777.1 | 1 | 396 | AAH11777.1 | |
| BC049220.1 | 1 | 403 | AAH49220.1 | |
| BC053350.1 | 1 | 387 | AAH53350.1 | |
| BP347679.1 | 1 | 415 | ||
| CX761192.1 | 1 | 427 |
More Information
Set Membership: |
|

Genomic Map