Assay Details
Target Gene Details
Entrez Gene ID: | 4731 |
Gene Name: | NADH:ubiquinone oxidoreductase subunit V3 |
Gene Aliases: |
CI-10k, CI-9KD |
Location: |
Chr.21:42893268-42911396 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFV3 | NM_001001503.1 | 2 | 167 | NP_001001503.1 |
| NM_021075.3 | 2 | 167 | NP_066553.3 | |
| XM_011529586.2 | 2 | 164 | XP_011527888.1 | |
| XM_017028359.1 | 2 | 162 | XP_016883848.1 | |
| AK056041.1 | 4 | 624 | BAB71080.1 | |
| AK289586.1 | 2 | 121 | ||
| AK308228.1 | 2 | 124 | ||
| AY173949.1 | 1 | 62 | AAO49719.1 | |
| BC021217.2 | 2 | 154 | AAH21217.2 | |
| BC033766.1 | 2 | 125 | AAH33766.1 | |
| BC054016.1 | 2 | 168 | AAH54016.1 | |
| BF311986.1 | 2 | 112 | ||
| BI829092.1 | 2 | 170 | ||
| CR542174.1 | 2 | 101 | CAG46971.1 | |
| CR542190.1 | 2 | 101 | CAG46987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587657 | Chr.21:42752699 - 43087910 on Build GRCh38 | Gain |
|
| esv3568210 | Chr.21:42868799 - 42962716 on Build GRCh38 | Loss |
|
| esv2666871 | Chr.21:42615442 - 43467915 on Build GRCh38 | Deletion |
|
| esv3584478 | Chr.21:42827914 - 42993457 on Build GRCh38 | Gain |
|
| nsv834105 | Chr.21:42873719 - 43030219 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map