Assay Details
Target Gene Details
Entrez Gene ID: | 570 |
Gene Name: | bile acid-CoA:amino acid N-acyltransferase |
Gene Aliases: |
BACAT, BAT |
Location: |
Chr.9:101360417-101385005 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAAT | NM_001127610.1 | NP_001121082.1 | ||
| NM_001701.3 | NP_001692.1 | |||
| AK315347.1 | ||||
| BC009567.1 | AAH09567.1 | |||
| BC107424.1 | AAI07425.1 | |||
| BG565737.1 | ||||
| CR541918.1 | CAG46716.1 | |||
| L34081.1 | AAC37550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037157 | Chr.9:101283479 - 101386951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map