Assay Details
Target Gene Details
Entrez Gene ID: | 51303 |
Gene Name: | FK506 binding protein 11 |
Gene Aliases: |
FKBP19 |
Location: |
Chr.12:48921959-48925547 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP11 | NM_001143781.1 | NP_001137253.1 | ||
| NM_001143782.1 | NP_001137254.1 | |||
| NM_016594.2 | NP_057678.1 | |||
| AF238079.1 | AAF63478.1 | |||
| AK094876.1 | ||||
| AK301455.1 | ||||
| AY358998.1 | AAQ89357.1 | |||
| BC027973.1 | AAH27973.1 | |||
| BC048292.1 | AAH48292.1 | |||
| BE857354.1 | ||||
| DA378205.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832404 | Chr.12:48864777 - 49064772 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map