Assay Details
Target Gene Details
Entrez Gene ID: | 55503 |
Gene Name: | transient receptor potential cation channel subfamily V member 6 |
Gene Aliases: |
ABP/ZF, CAT1, CATL, ECAC2, HSA277909, LP6728, ZFAB |
Location: |
Chr.7:142871203-142885745 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPV6 | NM_018646.5 | 13 | 1889 | NP_061116.5 |
| AF304463.1 | 13 | 1660 | AAG41951.1 | |
| AF365927.1 | 13 | 1876 | AAL40230.1 | |
| AF365928.1 | 9 | 1303 | AAM00356.1 | |
| AJ243500.2 | 13 | 1860 | CAC20416.2 | |
| AJ243501.2 | 13 | 1860 | CAC20417.2 | |
| AJ277909.1 | 3 | 381 | CAB92352.1 | |
| AJ487964.2 | 14 | 1829 | CAD32311.1 | |
| AK075103.1 | 7 | 1390 | BAC11402.1 | |
| AK075206.1 | 3 | 259 | BAC11471.1 | |
| AK291707.1 | 13 | 1890 | ||
| AY203947.1 | 10 | 2070 | AAP34470.1 | |
| KF534785.1 | 13 | 1889 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831168 | Chr.7:142830591 - 143025918 on Build GRCh38 | Gain |
|
| nsv949965 | Chr.7:142862343 - 142878755 on Build GRCh38 | Deletion |
|
| esv2759569 | Chr.7:142434958 - 143027060 on Build GRCh38 | Loss |
|
| nsv526015 | Chr.7:142864981 - 142944120 on Build GRCh38 | Gain |
|
| nsv520735 | Chr.7:142865498 - 142944120 on Build GRCh38 | Loss |
|
| nsv428187 | Chr.7:142618692 - 142909521 on Build GRCh38 | Loss |
|
| nsv1030700 | Chr.7:142867398 - 143010429 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map