Assay Details
Target Gene Details
Entrez Gene ID: | 51149 |
Gene Name: | chromosome 5 open reading frame 45 |
Gene Aliases: |
- |
Location: |
Chr.5:179837266-179858840 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C5orf45 | NM_001017987.2 | 5 | 795 | NP_001017987.1 |
| NM_016175.3 | 7 | 960 | NP_057259.2 | |
| AF153685.1 | 3 | 581 | AAD51611.1 | |
| AK293901.1 | ||||
| AK298954.1 | ||||
| BC002739.2 | 3 | 507 | AAH02739.2 | |
| BC069051.1 | 7 | 928 | AAH69051.1 | |
| BQ642639.1 | 5 | 751 | ||
| BX537968.1 | CAD97932.1 | |||
| BX647375.1 | 6 | 3914 | ||
| CR936785.1 | 6 | 2092 |
Target Gene Details
Entrez Gene ID: | 8878 |
Gene Name: | sequestosome 1 |
Gene Aliases: |
A170, FTDALS3, OSIL, PDB3, ZIP3, p60, p62, p62B |
Location: |
Chr.5:179806388-179838078 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SQSTM1 | NM_001142298.1 | 9 | 2345 | NP_001135770.1 |
| NM_001142299.1 | 9 | 2262 | NP_001135771.1 | |
| NM_003900.4 | 8 | 2332 | NP_003891.1 | |
| BC017222.1 | 8 | 2277 | AAH17222.1 | |
| BU625974.1 | 1 | 587 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469579 | Chr.5:179757830 - 179933191 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map