Assay Details
Target Gene Details
Entrez Gene ID: | 136242 |
Gene Name: | protease, serine 37 |
Gene Aliases: |
TRYX2 |
Location: |
Chr.7:141836278-141849725 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS37 | NM_001008270.2 | NP_001008271.2 | ||
| NM_001171951.1 | NP_001165422.1 | |||
| XM_005249946.4 | XP_005250003.1 | |||
| XM_005249947.3 | XP_005250004.1 | |||
| XM_011515792.2 | XP_011514094.1 | |||
| XM_011515793.2 | XP_011514095.1 | |||
| AA628371.1 | ||||
| AK097648.1 | ||||
| AK172866.1 | ||||
| BC137353.1 | ||||
| BC137354.1 | ||||
| BC144605.1 |
More Information
Set Membership: |
|

Genomic Map