Assay Details
Target Gene Details
Entrez Gene ID: | 23163 |
Gene Name: | golgi associated, gamma adaptin ear containing, ARF binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.17:75225464-75262393 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GGA3 | NM_001172703.2 | NP_001166174.1 | ||
| NM_001172704.2 | NP_001166175.1 | |||
| XM_017024388.1 | XP_016879877.1 | |||
| XM_017024389.1 | XP_016879878.1 | |||
| XM_017024390.1 | XP_016879879.1 | |||
| AK301895.1 | ||||
| AK302278.1 | ||||
| HY030361.1 |
Target Gene Details
Entrez Gene ID: | 51081 |
Gene Name: | mitochondrial ribosomal protein S7 |
Gene Aliases: |
MRP-S, MRP-S7, RP-S7, RPMS7, S7mt, bMRP27a |
Location: |
Chr.17:75261668-75266376 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS7 | NM_015971.3 | 1 | 128 | NP_057055.2 |
| AF077042.1 | 1 | 77 | AAD27775.1 | |
| AK222995.1 | 1 | 92 | BAD96715.1 | |
| BC000241.1 | 1 | 87 | AAH00241.1 | |
| BP379038.1 | 1 | 128 | ||
| BU599633.1 | 1 | 106 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510727 | Chr.17:75260216 - 75331456 on Build GRCh38 | Deletion |
|
| nsv1066312 | Chr.17:75260266 - 75323694 on Build GRCh38 | Gain |
|
| nsv576027 | Chr.17:75249939 - 75750289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map