Assay Details
Target Gene Details
Entrez Gene ID: | 50624 |
Gene Name: | CUB and zona pellucida like domains 1 |
Gene Aliases: |
ERG-1, ERG1, ITMAP1, UO-44 |
Location: |
Chr.10:122832155-122846175 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CUZD1 | NM_022034.5 | 9 | 2128 | NP_071317.2 |
| NR_037912.1 | 8 | 1977 | ||
| AF305835.1 | 6 | 1915 | AAG23215.1 | |
| AK289445.1 | 8 | 1659 | ||
| AY260047.1 | 9 | 2128 | AAP15458.1 | |
| AY260048.1 | 8 | 1828 | AAP15459.1 | |
| AY260049.1 | 8 | 1977 | AAP15460.1 | |
| AY260050.1 | 4 | 1220 | AAP15461.1 | |
| BC136755.1 | 9 | 1966 | ||
| BX538283.1 | 6 | 1452 | CAD98079.2 | |
| BX538284.1 | 6 | 1505 | CAD98080.2 | |
| BX648572.1 | 9 | 1810 |
Target Gene Details
Entrez Gene ID: | 100533195 |
Gene Name: | FAM24B-CUZD1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.10:122832155-122879641 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM24B-CUZD1 | NR_037915.1 | 11 | 2472 | |
| AK300849.1 | 11 | 2461 | ||
| AY358556.1 | 11 | 2759 | AAQ88920.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832010 | Chr.10:122826964 - 123037874 on Build GRCh38 | Gain |
|
| nsv1159779 | Chr.10:122823567 - 122836418 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map