Assay Details
Target Gene Details
Entrez Gene ID: | 51317 |
Gene Name: | PHD finger protein 21A |
Gene Aliases: |
BHC80, BM-006 |
Location: |
Chr.11:45929319-46121434 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv523952 | Chr.11:45961834 - 46006730 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map