Assay Details
Target Gene Details
Entrez Gene ID: | 6945 |
Gene Name: | MLX, MAX dimerization protein |
Gene Aliases: |
MAD7, MXD7, TCFL4, bHLHd13 |
Location: |
Chr.17:42567060-42573203 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLX | NM_170607.2 | NP_733752.1 | ||
| NM_198204.1 | NP_937847.1 | |||
| NM_198205.1 | NP_937848.1 | |||
| XM_017024991.1 | XP_016880480.1 | |||
| AF203978.1 | AAF14638.1 | |||
| AF213666.1 | AAG40145.1 | |||
| AF213667.1 | AAG40146.1 | |||
| AF213668.1 | AAG40147.1 | |||
| AK000150.1 | BAA90977.1 | |||
| AK290258.1 | ||||
| AK296114.1 | ||||
| AK303221.1 | ||||
| AK309628.1 | 4 | 299 | ||
| AK314378.1 | ||||
| AK315432.1 | ||||
| BC010689.2 | AAH10689.1 | |||
| BE907687.1 | ||||
| BT009812.1 | AAP88814.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064012 | Chr.17:42427786 - 42606703 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map