Assay Details
Target Gene Details
Entrez Gene ID: | 4343 |
Gene Name: | Mov10 RISC complex RNA helicase |
Gene Aliases: |
fSAP113, gb110 |
Location: |
Chr.1:112674312-112702377 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOV10 | XM_005270869.4 | 22 | 4365 | XP_005270926.1 |
| XM_005270870.4 | 22 | 4381 | XP_005270927.1 | |
| XM_017001318.1 | 22 | 4972 | XP_016856807.1 |
Target Gene Details
Entrez Gene ID: | 389 |
Gene Name: | ras homolog family member C |
Gene Aliases: |
ARH9, ARHC, H9, RHOH9 |
Location: |
Chr.1:112701127-112707403 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHOC | NM_001042678.1 | 5 | 1045 | NP_001036143.1 |
| NM_001042679.1 | 6 | 1012 | NP_001036144.1 | |
| NM_175744.4 | 6 | 1114 | NP_786886.1 | |
| AK094474.1 | 6 | 1922 | ||
| BC007245.1 | 6 | 809 | AAH07245.1 | |
| BC009177.2 | 6 | 809 | AAH09177.1 | |
| BC052808.1 | 4 | 734 | AAH52808.1 | |
| BX648784.1 | 5 | 742 | ||
| CA307508.1 | 1 | 323 | ||
| CB851337.1 | 1 | 302 | ||
| L25081.1 | 5 | 788 | AAC33179.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv27061 | Chr.1:112150007 - 112703641 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map