Assay Details
Target Gene Details
Entrez Gene ID: | 56649 |
Gene Name: | transmembrane protease, serine 4 |
Gene Aliases: |
CAPH2, MT-SP2, TMPRSS3 |
Location: |
Chr.11:118077012-118125505 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS4 | NM_001083947.1 | NP_001077416.1 | ||
| NM_001173551.1 | NP_001167022.1 | |||
| NM_001173552.1 | NP_001167023.1 | |||
| NM_001290094.1 | NP_001277023.1 | |||
| NM_001290096.1 | NP_001277025.1 | |||
| NM_019894.3 | NP_063947.1 | |||
| NR_110734.1 | ||||
| XM_005271613.3 | XP_005271670.1 | |||
| XM_005271614.2 | XP_005271671.1 | |||
| XM_005271615.3 | XP_005271672.1 | |||
| XM_011542901.2 | XP_011541203.1 | |||
| XM_011542902.2 | XP_011541204.1 | |||
| XM_011542903.2 | XP_011541205.1 | |||
| XM_011542904.2 | XP_011541206.1 | |||
| AF179224.1 | AAF74526.1 | |||
| AF216312.1 | AAF31436.1 | |||
| AK172766.1 | BAD18749.1 | |||
| AK290361.1 | ||||
| AK296855.1 | ||||
| AK303173.1 | ||||
| AK304113.1 | ||||
| AK304215.1 | ||||
| AK308434.1 | ||||
| AY358530.1 | AAQ88894.1 | |||
| BC011703.2 | AAH11703.1 | |||
| BP220493.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556464 | Chr.11:118076983 - 118161527 on Build GRCh38 | Gain |
|
| nsv1049373 | Chr.11:118066351 - 118134273 on Build GRCh38 | Loss |
|
| esv3892103 | Chr.11:118081382 - 118161527 on Build GRCh38 | Gain |
|
| nsv1159938 | Chr.11:118075976 - 118157452 on Build GRCh38 | Duplication |
|
| nsv509444 | Chr.11:118033438 - 118117536 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map