Assay Details
Target Gene Details
Entrez Gene ID: | 144100 |
Gene Name: | pleckstrin homology domain containing A7 |
Gene Aliases: |
- |
Location: |
Chr.11:16787153-17014423 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLEKHA7 | NM_175058.4 | 23 | 3841 | NP_778228.3 |
| XM_017017240.1 | XP_016872729.1 | |||
| XM_017017241.1 | 25 | 4052 | XP_016872730.1 | |
| XM_017017242.1 | 24 | 3932 | XP_016872731.1 | |
| XM_017017243.1 | 23 | 3851 | XP_016872732.1 | |
| BC033239.1 | 14 | 2721 | AAH33239.1 | |
| BC041452.1 | 5 | 1179 | AAH41452.1 | |
| BC071599.1 | 23 | 3837 | AAH71599.1 | |
| BX648885.1 | 2 | 2624 |
More Information
Additional Information:
For this assay, SNP(s) [rs574577759] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map