Assay Details
Target Gene Details
Entrez Gene ID: | 83417 |
Gene Name: | Fc receptor like 4 |
Gene Aliases: |
CD307d, FCRH4, IGFP2, IRTA1 |
Location: |
Chr.1:157573749-157598080 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FCRL4 | NM_031282.2 | 4 | 613 | NP_112572.1 |
| XM_011510034.1 | 4 | 589 | XP_011508336.1 | |
| AF329490.1 | 4 | 477 | AAL23900.1 | |
| AF329492.1 | 2 | 218 | AAL23902.1 | |
| AF343659.1 | 4 | 613 | AAK31322.1 | |
| AF343660.1 | 4 | 613 | AAK31323.1 | |
| AF343661.1 | 4 | 613 | AAK31324.1 | |
| AF397452.1 | 4 | 477 | AAK93970.1 | |
| BC125173.1 | 4 | 571 | ||
| BC125174.1 | 4 | 571 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2718507 | Chr.1:157533583 - 157635387 on Build GRCh38 | Deletion |
|
| esv3587653 | Chr.1:157587056 - 157588902 on Build GRCh38 | Loss |
|
| esv2661330 | Chr.1:157587008 - 157588897 on Build GRCh38 | Deletion |
|
| nsv3154 | Chr.1:157562046 - 157596752 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map