Assay Details
Target Gene Details
Entrez Gene ID: | 2138 |
Gene Name: | EYA transcriptional coactivator and phosphatase 1 |
Gene Aliases: |
BOP, BOR, BOS1, OFC1 |
Location: |
Chr.8:71197433-71548130 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EYA1 | NM_000503.5 | 18 | 3270 | NP_000494.2 |
| NM_001288574.1 | 18 | 3252 | NP_001275503.1 | |
| NM_001288575.1 | 18 | 3188 | NP_001275504.1 | |
| NM_172058.3 | 17 | 2862 | NP_742055.1 | |
| NM_172059.3 | 16 | 2579 | NP_742056.1 | |
| NM_172060.3 | 16 | 2734 | NP_742057.1 | |
| XM_011517483.2 | 17 | 7567 | XP_011515785.1 | |
| XM_011517484.2 | 19 | 3013 | XP_011515786.2 | |
| XM_017013201.1 | 19 | 3056 | XP_016868690.1 | |
| XM_017013202.1 | 19 | 2900 | XP_016868691.1 | |
| XM_017013203.1 | 19 | 3025 | XP_016868692.1 | |
| XM_017013204.1 | 19 | 3011 | XP_016868693.1 | |
| XM_017013206.1 | 20 | 3009 | XP_016868695.1 | |
| XM_017013207.1 | 18 | 2937 | XP_016868696.1 | |
| XM_017013208.1 | 18 | 3292 | XP_016868697.1 | |
| XM_017013209.1 | 18 | 2812 | XP_016868698.1 | |
| XM_017013210.1 | 18 | 2921 | XP_016868699.1 | |
| XM_017013211.1 | 18 | 2900 | XP_016868700.1 | |
| XM_017013212.1 | 17 | 3167 | XP_016868701.1 | |
| XM_017013213.1 | 17 | 7705 | XP_016868702.1 | |
| AJ000097.1 | 17 | 2807 | CAA03922.1 | |
| AJ000098.1 | 18 | 3268 | CAA03923.1 | |
| AK127798.1 | 18 | 2688 | ||
| BQ223588.1 | 1 | 230 |
More Information
Set Membership: |
|

Genomic Map