Assay Details
Target Gene Details
Entrez Gene ID: | 5228 |
Gene Name: | placental growth factor |
Gene Aliases: |
D12S1900, PGFL, PIGF, PLGF, PlGF-2, SHGC-10760 |
Location: |
Chr.14:74941830-74955764 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGF | NM_001207012.1 | 6 | 1500 | NP_001193941.1 |
| NM_001293643.1 | 7 | 1560 | NP_001280572.1 | |
| NM_002632.5 | 7 | 1563 | NP_002623.2 | |
| A18411.1 | 6 | 1299 | CAA01393.1 | |
| BC001422.2 | 7 | 1387 | AAH01422.1 | |
| BC007255.1 | 7 | 1368 | AAH07255.1 | |
| BC007789.2 | 7 | 1365 | AAH07789.1 | |
| BX356823.2 | ||||
| X54936.1 | 6 | 1299 | CAA38698.1 |
More Information
Set Membership: |
|

Genomic Map