Assay Details
Target Gene Details
Entrez Gene ID: | 54407 |
Gene Name: | solute carrier family 38 member 2 |
Gene Aliases: |
ATA2, PRO1068, SAT2, SNAT2 |
Location: |
Chr.12:46358188-46372862 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC38A2 | NM_001307936.1 | NP_001294865.1 | ||
| NM_018976.4 | NP_061849.2 | |||
| AB037803.2 | BAA92620.2 | |||
| AF259799.1 | AAK38510.1 | |||
| AF298897.1 | AAG24618.1 | |||
| AJ344099.1 | CAC51434.1 | |||
| AK001700.1 | BAA91846.1 | |||
| AK172784.1 | BAD18765.1 | |||
| AK291644.1 | ||||
| AK293858.1 | ||||
| BC040342.1 | AAH40342.1 | |||
| CR457267.1 | CAG33548.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832398 | Chr.12:46217909 - 46431747 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map