Assay Details
Target Gene Details
Entrez Gene ID: | 1737 |
Gene Name: | dihydrolipoamide S-acetyltransferase |
Gene Aliases: |
DLTA, PDC-E2, PDCE2 |
Location: |
Chr.11:112024814-112064278 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLAT | NM_001931.4 | 9 | 1908 | NP_001922.2 |
| XM_011542647.2 | 9 | 1555 | XP_011540949.1 | |
| AK057299.1 | 9 | 1908 | ||
| AK223596.1 | 9 | 1297 | BAD97316.1 | |
| AK296274.1 | 9 | 1187 | ||
| AK297103.1 | 6 | 654 | ||
| AK299562.1 | 7 | 745 | ||
| AK303930.1 | 10 | 1133 | ||
| AL832703.1 | 9 | 1279 | ||
| BC039084.1 | 9 | 1277 | AAH39084.1 | |
| X13822.1 | 3 | 241 | CAA32052.1 | |
| Y00978.1 | 9 | 1153 | CAA68787.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map