Assay Details
Target Gene Details
Entrez Gene ID: | 29899 |
Gene Name: | G-protein signaling modulator 2 |
Gene Aliases: |
CMCS, DFNB82, LGN, PINS |
Location: |
Chr.1:108875350-108934335 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPSM2 | NM_001321038.1 | 5 | 722 | NP_001307967.1 |
| NM_001321039.1 | 5 | 954 | NP_001307968.1 | |
| NM_013296.4 | 5 | 954 | NP_037428.3 | |
| XM_006710589.2 | 4 | 1413 | XP_006710652.1 | |
| XM_011541301.2 | 6 | 1231 | XP_011539603.1 | |
| XM_011541302.2 | 6 | 1893 | XP_011539604.1 | |
| XM_011541303.2 | 5 | 1649 | XP_011539605.1 | |
| XM_017001097.1 | 6 | 1799 | XP_016856586.1 | |
| XM_017001098.1 | 6 | 1567 | XP_016856587.1 | |
| AB445462.1 | 5 | 954 | ||
| AK295563.1 | 5 | 541 | ||
| AK314249.1 | 5 | 950 | ||
| AY136740.1 | 4 | 437 | AAN01266.1 | |
| BC027732.2 | 5 | 730 | AAH27732.1 | |
| CR456786.1 | 4 | 437 | CAG33067.1 | |
| KX085485.1 | 5 | 954 | ||
| U54999.1 | 4 | 610 | AAB40385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1005738 | Chr.1:108868275 - 108917679 on Build GRCh38 | Loss |
|
| dgv15n21 | Chr.1:108873603 - 108941145 on Build GRCh38 | Gain |
|
| esv3587141 | Chr.1:108866483 - 108915393 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map