Assay Details
Target Gene Details
Entrez Gene ID: | 81888 |
Gene Name: | hydroxypyruvate isomerase (putative) |
Gene Aliases: |
HT036 |
Location: |
Chr.1:43450607-43454241 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HYI | XM_006710937.3 | 9 | 1463 | XP_006711000.1 |
Target Gene Details
Entrez Gene ID: | 23334 |
Gene Name: | seizure threshold 2 homolog (mouse) |
Gene Aliases: |
C1orf84, EIEE18, KIAA0467, SZT2A, SZT2B |
Location: |
Chr.1:43389884-43454247 on Build GRCh38 |
Assay Gene Location: | Within Exon 75 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SZT2 | NM_015284.3 | 71 | 10606 | NP_056099.3 |
| XM_005270686.3 | 73 | 10838 | XP_005270743.1 | |
| XM_006710501.3 | 72 | 10725 | XP_006710564.1 | |
| XM_011541106.2 | 72 | 10876 | XP_011539408.1 | |
| XM_011541107.2 | 69 | 10373 | XP_011539409.1 | |
| XM_017000819.1 | 73 | 10685 | XP_016856308.1 | |
| XM_017000820.1 | 72 | 10667 | XP_016856309.1 | |
| XM_017000821.1 | 69 | 10359 | XP_016856310.1 | |
| AB007936.2 | 57 | 8458 | BAA32312.2 | |
| AK001249.2 | 6 | 1417 | ||
| AK027078.1 | 12 | 2151 | BAB15649.1 | |
| AK126972.1 | 10 | 2508 | BAC86771.1 | |
| AL117402.1 | 6 | 1400 | CAB55903.1 | |
| BC041069.1 | 13 | 2287 | ||
| BC052802.1 | 14 | 2301 | AAH52802.1 | |
| BC082968.1 | 14 | 2306 | AAH82968.1 | |
| BC151232.1 | 57 | 8458 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527798 | Chr.1:43446919 - 43451738 on Build GRCh38 | Loss |
|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
|
| nsv950701 | Chr.1:43418430 - 43453929 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map