Assay Details
Target Gene Details
Entrez Gene ID: | 8751 |
Gene Name: | ADAM metallopeptidase domain 15 |
Gene Aliases: |
MDC15 |
Location: |
Chr.1:155051272-155062776 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
Target Gene Details
Entrez Gene ID: | 100505666 |
Gene Name: | uncharacterized LOC100505666 |
Gene Aliases: |
- |
Location: |
Chr.1:155045191-155063991 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100505666 | NR_040773.1 | |||
| BM047608.1 | ||||
| DB152307.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524157 | Chr.1:155054466 - 155058081 on Build GRCh38 | Loss |
|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| esv3587616 | Chr.1:155055926 - 155098657 on Build GRCh38 | Gain |
|
| esv3587615 | Chr.1:155047853 - 155175013 on Build GRCh38 | Gain |
|
| nsv952212 | Chr.1:154998225 - 155087624 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map