Assay Details
Target Gene Details
Entrez Gene ID: | 128869 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class U |
Gene Aliases: |
CDC91L1, GAB1 |
Location: |
Chr.20:34560542-34677285 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGU | NM_080476.4 | 12 | 1633 | NP_536724.1 |
| XM_011528542.2 | 6 | 924 | XP_011526844.1 | |
| XM_017027664.1 | 11 | 1307 | XP_016883153.1 | |
| AB086842.1 | 12 | 1439 | BAC53626.1 | |
| AK075507.1 | 12 | 1458 | BAC11660.1 | |
| AK095044.1 | 1 | 2928 | ||
| AK095356.1 | 1 | 3618 | ||
| AY358816.1 | 12 | 1463 | AAQ89175.1 | |
| AY422169.1 | 12 | 1633 | AAR23798.1 | |
| BC030512.1 | 12 | 1444 | AAH30512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3359 | Chr.20:34531574 - 34576232 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map