Assay Details
Target Gene Details
Entrez Gene ID: | 116931 |
Gene Name: | mediator complex subunit 12 like |
Gene Aliases: |
NOPAR, TNRC11L, TRALP, TRALPUSH |
Location: |
Chr.3:151085665-151436677 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED12L | NM_053002.5 | 1 | 111 | NP_443728.3 |
| XM_006713487.3 | 2 | 383 | XP_006713550.1 | |
| XM_011512390.2 | 2 | 383 | XP_011510692.1 | |
| XM_011512394.2 | 2 | 383 | XP_011510696.1 | |
| XM_011512399.2 | 2 | 383 | XP_011510701.1 | |
| XM_017005676.1 | 2 | 383 | XP_016861165.1 | |
| XM_017005677.1 | 2 | 383 | XP_016861166.1 | |
| XM_017005678.1 | 2 | 383 | XP_016861167.1 | |
| XM_017005679.1 | 2 | 383 | XP_016861168.1 | |
| XM_017005680.1 | 2 | 383 | XP_016861169.1 | |
| AF388364.1 | 1 | 515 | AAL09579.1 | |
| AF388365.1 | 1 | 515 | AAL09580.1 | |
| AF399708.1 | AAO38813.1 | |||
| AK128165.1 | 2 | 384 | ||
| BC144559.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759188 | Chr.3:151073838 - 151239697 on Build GRCh38 | Gain |
|
| nsv829756 | Chr.3:150974849 - 151171951 on Build GRCh38 | Gain+Loss |
|
| nsv592045 | Chr.3:151052827 - 151377305 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map