Assay Details
Target Gene Details
Entrez Gene ID: | 27005 |
Gene Name: | ubiquitin specific peptidase 21 |
Gene Aliases: |
USP16, USP23 |
Location: |
Chr.1:161159458-161165726 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP21 | NM_001014443.2 | NP_001014443.1 | ||
| NM_001319848.1 | NP_001306777.1 | |||
| NM_012475.4 | NP_036607.3 | |||
| XM_011509414.1 | XP_011507716.1 | |||
| XM_017001036.1 | XP_016856525.1 | |||
| XM_017001037.1 | XP_016856526.1 | |||
| XM_017001038.1 | XP_016856527.1 | |||
| XM_017001039.1 | XP_016856528.1 | |||
| XM_017001040.1 | XP_016856529.1 | |||
| XM_017001041.1 | XP_016856530.1 | |||
| XM_017001042.1 | XP_016856531.1 | |||
| AB208899.1 | BAD92136.1 | |||
| AF177758.1 | AAD54321.1 | |||
| AF217979.1 | AAG17222.1 | |||
| AF233442.1 | AAF61308.1 | |||
| AK292319.1 | ||||
| AK310326.1 | ||||
| AL157417.1 | CAB75649.1 | |||
| BC003130.1 | AAH03130.2 | |||
| BC090946.1 | AAH90946.1 | |||
| BC136291.1 |
More Information
Set Membership: |
|

Genomic Map