Assay Details
Target Gene Details
Entrez Gene ID: | 1610 |
Gene Name: | D-amino acid oxidase |
Gene Aliases: |
DAAO, DAMOX, OXDA |
Location: |
Chr.12:108880073-108901043 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DAO | NM_001917.4 | NP_001908.3 | ||
| XM_005268692.3 | XP_005268749.1 | |||
| XM_011538004.2 | XP_011536306.1 | |||
| XM_011538005.2 | XP_011536307.1 | |||
| AK312995.1 | ||||
| AX741655.1 | CAD90933.1 | |||
| AX741656.1 | CAD90934.1 | |||
| AX741658.1 | CAD90936.1 | |||
| AX741659.1 | CAD90937.1 | |||
| AX776478.1 | CAE11742.1 | |||
| AX776479.1 | CAE11743.1 | |||
| AX776480.1 | CAE11744.1 | |||
| AX786386.1 | CAE11959.1 | |||
| BC029057.1 | AAH29057.1 | |||
| BC074770.2 | AAH74770.1 | |||
| BX538210.1 | CAD98069.1 | |||
| DA085245.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524524 | Chr.12:108878275 - 108899544 on Build GRCh38 | Gain |
|
| esv2422361 | Chr.12:108792678 - 109118360 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map