Assay Details
Target Gene Details
Entrez Gene ID: | 29964 |
Gene Name: | prickle planar cell polarity protein 4 |
Gene Aliases: |
C6orf49, OBTP, OEBT, TOMM6 |
Location: |
Chr.6:41780762-41787372 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRICKLE4 | NM_013397.5 | NP_037529.3 | ||
| BC110459.1 | AAI10460.1 |
Target Gene Details
Entrez Gene ID: | 100188893 |
Gene Name: | translocase of outer mitochondrial membrane 6 |
Gene Aliases: |
OBTP, TOM6 |
Location: |
Chr.6:41787443-41789896 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOMM6 | NM_001134493.1 | NP_001127965.1 | ||
| AJ420506.1 | 1 | 692 | ||
| BM783950.1 | 1 | 93 |
More Information
Set Membership: |
|

Genomic Map