Assay Details
Target Gene Details
Entrez Gene ID: | 26168 |
Gene Name: | SUMO1/sentrin/SMT3 specific peptidase 3 |
Gene Aliases: |
SMT3IP1, SSP3, Ulp1 |
Location: |
Chr.17:7561992-7571969 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP3 | NM_015670.5 | NP_056485.2 | ||
| AF199459.1 | AAL25652.1 | |||
| AK000923.1 | 2 | 1016 | ||
| AL050283.1 | CAB43384.2 | |||
| AL834294.1 | CAD38967.1 | |||
| AY008763.1 | AAG33252.1 | |||
| BC048306.1 | AAH48306.1 | |||
| BC080658.1 | AAH80658.1 |
Target Gene Details
Entrez Gene ID: | 100533955 |
Gene Name: | SENP3-EIF4A1 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.17:7561992-7579006 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP3-EIF4A1 | NR_037926.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv457659 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map