Assay Details
Target Gene Details
Entrez Gene ID: | 58529 |
Gene Name: | myozenin 1 |
Gene Aliases: |
CS-2, FATZ, MYOZ |
Location: |
Chr.10:73631612-73641757 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYOZ1 | NM_021245.3 | 1 | 160 | NP_067068.1 |
| AJ278124.1 | 1 | 153 | CAB92967.1 | |
| BC025753.1 | 1 | 160 | AAH25753.1 |
More Information
Set Membership: |
|

Genomic Map