Assay Details
Target Gene Details
Entrez Gene ID: | 116519 |
Gene Name: | apolipoprotein A5 |
Gene Aliases: |
APOAV, RAP3 |
Location: |
Chr.11:116789370-116792420 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOA5 | NM_001166598.1 | 4 | 946 | NP_001160070.1 |
| NM_052968.4 | 4 | 970 | NP_443200.2 | |
| XM_017017169.1 | 3 | 1428 | XP_016872658.1 | |
| AF202889.1 | 3 | 905 | AAF25661.1 | |
| AF202890.1 | 3 | 905 | AAF25662.1 | |
| AK292739.1 | 3 | 917 | ||
| AK308047.1 | 2 | 1407 | ||
| AK315229.1 | 3 | 917 | ||
| AY358749.1 | 3 | 908 | AAQ89109.1 | |
| BC101787.1 | 3 | 933 | AAI01788.1 | |
| BC101789.1 | 3 | 933 | AAI01790.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv507 | Chr.11:116763503 - 116804070 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map