Assay Details
Target Gene Details
Entrez Gene ID: | 1289 |
Gene Name: | collagen type V alpha 1 |
Gene Aliases: |
EDSC |
Location: |
Chr.9:134641790-134844843 on Build GRCh38 |
Assay Gene Location: | Within Exon 68 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL5A1 | NM_000093.4 | 66 | 6899 | NP_000084.3 |
| NM_001278074.1 | 66 | 6899 | NP_001265003.1 | |
| AB209864.1 | 65 | 6378 | BAD93101.1 | |
| AB371583.1 | 66 | 6899 | ||
| AK122763.1 | 1 | 186 |
Target Gene Details
Entrez Gene ID: | 101448202 |
Gene Name: | uncharacterized LOC101448202 |
Gene Aliases: |
- |
Location: |
Chr.9:134819415-134872618 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101448202 | NR_103451.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv1043726 | Chr.9:134566621 - 135388584 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map