Assay Details
Target Gene Details
Entrez Gene ID: | 6472 |
Gene Name: | serine hydroxymethyltransferase 2 |
Gene Aliases: |
GLYA, HEL-S-51e, SHMT |
Location: |
Chr.12:57229573-57234935 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv952189 | Chr.12:57127118 - 57245617 on Build GRCh38 | Deletion |
|
| nsv519293 | Chr.12:57189889 - 57243810 on Build GRCh38 | Loss |
|
| dgv96n21 | Chr.12:57139199 - 57243810 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map