Assay Details
Target Gene Details
Entrez Gene ID: | 58484 |
Gene Name: | NLR family CARD domain containing 4 |
Gene Aliases: |
AIFEC, CARD12, CLAN, CLAN1, CLANA, CLANB, CLANC, CLAND, CLR2.1, FCAS4, IPAF |
Location: |
Chr.2:32224449-32265743 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRC4 | NM_001199138.1 | NP_001186067.1 | ||
| NM_001199139.1 | NP_001186068.1 | |||
| NM_001302504.1 | NP_001289433.1 | |||
| NM_021209.4 | NP_067032.3 | |||
| XM_017004619.1 | XP_016860108.1 | |||
| AF376061.1 | AAK53443.1 | |||
| AK095467.1 | ||||
| AK292673.1 | ||||
| AK314762.1 | ||||
| AL389934.1 | CAB97523.1 | |||
| AY027787.1 | AAK14776.1 | |||
| AY027788.1 | AAK14777.1 | |||
| AY032589.1 | AAK38730.1 | |||
| AY035391.1 | AAK59843.1 | |||
| AY358152.1 | AAQ88519.1 | |||
| BC031555.1 | AAH31555.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3590243 | Chr.2:32217953 - 32258270 on Build GRCh38 | Gain |
|
| nsv2667 | Chr.2:32219741 - 32264686 on Build GRCh38 | Deletion |
|
| esv2422335 | Chr.2:32187261 - 32975514 on Build GRCh38 | Duplication |
|
| nsv833670 | Chr.2:32070709 - 32256788 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map