Assay Details
Target Gene Details
Entrez Gene ID: | 3973 |
Gene Name: | luteinizing hormone/choriogonadotropin receptor |
Gene Aliases: |
HHG, LCGR, LGR2, LH/CG-R, LH/CGR, LHR, LHRHR, LSH-R, ULG5 |
Location: |
Chr.2:48686774-48755741 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LHCGR | NM_000233.3 | NP_000224.2 | ||
| XM_011532828.1 | XP_011531130.1 | |||
| XM_011532831.1 | XP_011531133.1 | |||
| XM_017004089.1 | XP_016859578.1 | |||
| AK123498.1 | ||||
| DA376746.1 | ||||
| M63108.1 | AAA59515.1 | |||
| M73746.1 | AAA70231.1 | |||
| S57793.1 | AAB19917.2 |
Target Gene Details
Entrez Gene ID: | 286749 |
Gene Name: | STON1-GTF2A1L readthrough |
Gene Aliases: |
SALF |
Location: |
Chr.2:48529925-48776517 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STON1-GTF2A1L | NM_001198593.1 | NP_001185522.1 | ||
| AK309740.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1002793 | Chr.2:48544081 - 49239411 on Build GRCh38 | Gain |
|
| nsv833992 | Chr.2:48587084 - 48770898 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map