Assay Details
Target Gene Details
Entrez Gene ID: | 55152 |
Gene Name: | DALR anticodon binding domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.3:49015399-49021386 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DALRD3 | NM_018114.5 | NP_060584.3 | ||
| XM_006713219.2 | XP_006713282.1 | |||
| XM_011533892.1 | XP_011532194.1 | |||
| XM_017006723.1 | XP_016862212.1 | |||
| AK093204.1 | BAC04095.1 | |||
| AK093294.1 | BAC04123.1 | |||
| AK308273.1 | ||||
| HY047953.1 |
Target Gene Details
Entrez Gene ID: | 25915 |
Gene Name: | NADH:ubiquinone oxidoreductase complex assembly factor 3 |
Gene Aliases: |
2P1, C3orf60, E3-3 |
Location: |
Chr.3:49020475-49023495 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFAF3 | NM_199074.1 | NP_951056.1 | ||
| BQ652614.1 |
More Information
Set Membership: |
|

Genomic Map